missing translation for 'onlineSavingsMsg'
Learn More

Transglutaminase II Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ Transglutaminase II Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia MS-224-PABX

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12690597

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: CUB 7402

Isotype: IgG1/κ

Species Reactivity: Human, Dog, Rabbit and Guinea Pig. Does not react with mouse and rat. Others not known.

Epitope: aa 447-478

Immunogen: Purified guinea pig liver TGase II

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-1 inhibits the TGase II activity.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Affinity Purification
  • Biological Blockade

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Transglutaminase II Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Neuroscience
Canine, Guinea Pig, Human, Rabbit
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
CUB 7402
Unconjugated
Purified guinea pig liver TGase II
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales