Learn More
Synaptophysin Ab-4 Rabbit Polyclonal Antibody, Epredia™
Provide accurate, reproducible results Epredia™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.
Marca: Epredia RB-1461-P1ABX
Detalles adicionales : Peso : 1.00000kg
Descripción
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide from C-terminus of human synaptophysin
Molecular Weight: 38kDa
Positive Control: IMR-5 cells, Pancreas or Pheochromocytoma or GI tumor
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Especificaciones
Synaptophysin Ab-4 | |
Polyclonal | |
Unconjugated | |
A synthetic peptide from C-terminus of human synaptophysin | |
RUO | |
Primary | |
Concentrated |
Western Blot | |
1mg/mL | |
Rabbit | |
500 μL | |
Cancer and Tumor Biology | |
Human | |
IgG |