missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Survivin Rabbit Polyclonal Antibody with BSA and azide

Survivin, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Marca:  Epredia™ RB-9245-P1

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12693507

  • 342.00€ / 500 microlitros

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Survivin is a new inhibitor of apoptosis (IAP) protein. It is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. The overexpression of surviving in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.

Host Species: Rabbit

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human survivin

Molecular Weight: 16.5kDa

Positive Control: Normal pancreas (α-cells) or prostate CA

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Survivin
Polyclonal
Unconjugated
Rabbit
500μL
Apoptosis
Primary
Human, Murine
Immunohistochemistry (Paraffin)
200µg/mL
Concentrated
A synthetic peptide derived from the C-terminus of human survivin
RUO
Primary
Polyclonal
Videos
SDS
Documentos
Certificados