missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ RAD51 Ab-1 Mouse Monoclonal Antibody, biotin labeled with BSA and azide

Provide accurate, reproducible results with the Epredia™ RAD51 Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-988-P1ABX

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12682627

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Rad51 (a eukaryotic homologue of Escherichia coli RecA) is similar to RecA, both biochemically and structurally: It promotes homologous pairing and strand exchange within a regular nucleoprotein filament. In yeast, the Rad51 family of related proteins also includes Rad55, Rad57 and Dmc1. In mammalian cells, seven genes in this family have been identified (HsRAD51, XRCC2, XRCC3, RAD51B/hREC2 and HsDMC1, RAD51C, and RAD 51D). Recent studies suggested that Rad51 is involved not only in homologous recombination but also in cell proliferation regulation.

Host Species: Mouse

Clone: 51RAD01; same as 3C10

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant RAD51 protein

Molecular Weight: 37-43kDa

Positive Control: HeLa cells, Testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

RAD51 Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
DNA Repair
Human, Mouse, Rat
IgG2a κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
51RAD01; same as 3C10
Unconjugated
Recombinant Rad51 protein
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales