missing translation for 'onlineSavingsMsg'
Learn More

PDGFRα Ab-1 Rabbit Polyclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ PDGFRα Ab-1, Rabbit Polyclonal Antibody.

Marca:  Epredia RB-1691-P

Detalles adicionales : Peso : 0.00250kg

 Ver más versiones de este producto

Código de producto. 12693127

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and ß coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, alpha while PDGF BB can bind and activate to both PDGFR, α and β

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of the precursor form of human PDGFRα

Molecular Weight: ∽190kDa

Postitive Control: Ovarian CA

Cellular Localization: Membrane Specific for PDGFR, α and does not react with PDGFR, β.

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

PDGFR, alpha Ab-1
Polyclonal
1mg/mL
Rabbit
Purified with BSA and azide
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
A synthetic peptide derived from the C-terminus of the precursor form of human PDGFR, alpha
1 mL
Growth Factor
Human, Mouse
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales