missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p53 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Achieve accurate, reproducible results with the Epredia™ p53 Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-104-P

Detalles adicionales : Peso : 0.00100kg

 Ver más versiones de este producto

Código de producto. 12668706

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 240

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 212-217

Immunogen: Murine p53 protein corresponding to aa 14-289

Molecular Weight: 53kDa

Positive Control: MDA-231 or SKBR-3 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Methacarn or formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

p53 Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
Cell Cycle
Human
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
PAb 240
Unconjugated
Murine p53 protein corresponding to aa14-289
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales