missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ NFκB/p65 (Rel A) Rabbit Polyclonal Antibody with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ NFκB/p65 (Rel A), Rabbit Polyclonal Antibody.

Marca:  Epredia™ RB-9034-P

Detalles adicionales : Peso : 0.00100kg

 Ver más versiones de este producto

Código de producto. 12693367

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

NFκB is a heterodimer consisting of a 50kDa DNA binding subunit and a 65kDa transactivating subunit (RelA). The p50/p65 heterodimer is present in the cytoplasm in inactive form bound to its inhibitor, IκB. In unstimulated cells, nuclear localization signals present on p65 are masked by members of the IκB family of inhibitory proteins. Subsequent to cell stimulation, IκB undergoes phosphorylation, ubiquitination and degradation by a proteosome-dependent pathway, allowing nuclear translocation of the active dimeric NFκB transcription factor.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat and Rabbit. Others not tested.

Epitope: Internal domain

Immunogen: A synthetic peptide corresponding to internal domain of human NFκB/p65

Molecular Weight: 65kDa

Positive Control: Prostate gland and placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

NFkappaB/p65 (Rel A)
Polyclonal
200µg/mL
Rabbit
1 mL
Transcription Regulation
Human, Rabbit
IgG
Western Blot
Polyclonal
Unconjugated
A synthetic peptide corresponding to internal domain of human NFkappaB p65
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales