missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Negative Control for Rabbit IgG Ab-1 with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody.

Marca:  Epredia™ NC-100-P

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 12672657

  • 340.00€ / 1 ml

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This IgG was purified from serum collected from a rabbit prior to immunization. It does not react with any antigen of human or animal cells. The serum was purified under the same experimental conditions as for the other rabbit polyclonal antibodies of NeoMarkers. In order to serve as a true negative control, this IgG should be used under the identical conditions and at the same concentration as NeoMarkers' other purified rabbit polyclonal IgG antibodies.

Host Species: Rabbit

Isotype: IgG

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Negative Control for Rabbit IgG Ab-1
Polyclonal
1mg/mL
Concentrated; Purified with BSA and azide
IgG
RUO
Primary
Polyclonal
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
Rabbit
1mL
Negative Controls
Primary
All
Videos
SDS
Documentos
Certificados