missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Negative Control for Mouse IgG2b Ab-1 with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Mouse IgG2b Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ NC-1391-R7

Detalles adicionales : Peso : 0.00700kg

 Ver más versiones de este producto

Código de producto. 12602687

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

This mouse monoclonal IgG2b was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG2b does not react with any antigen of human or animal cells. The monoclonal IgG2b was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG2b should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG2b antibodies.

Host Species: Mouse

Clone: NCG2B.01

Isotype: IgG2b/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Negative Control for Mouse IgG2b Ab-1
Monoclonal
Unconjugated
7 mL
Negative Controls
All species
IgG2b κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
NCG2B.01
Mouse
RUO
Primary
Prediluted
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales