missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lab Vision™ Negative Control for Mouse IgG2b Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Negative Control for Mouse IgG2b Ab-1, Mouse Monoclonal Antibody.

Marca:  Thermo Scientific™ NC-1391-P0

Detalles adicionales : Peso : 0.00170kg

 Ver más versiones de este producto

Código de producto. 12652677

  • 101.98€ / 100 microlitros


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

This mouse monoclonal IgG2b was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG2b does not react with any antigen of human or animal cells. The monoclonal IgG2b was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG2b should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG2b antibodies.

Host Species: Mouse

Clone: NCG2B.01

Isotype: IgG2b/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Negative Control for Mouse IgG2b Ab-1
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
100μL
Negative Controls
Primary
All
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
NCG2B.01
Unconjugated
Concentrated
IgG2b kappa
RUO
Primary
Monoclonal
Videos
Documentos