missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ MMP-3 (Stromelysin-1) Ab-1 Mouse Monoclonal Antibody, biotin labeled with BSA and azide

Epredia™ MMP-3 (Stromelysin-1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia™ MS-809-B

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12692357

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »


Descripción

Descripción

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Stromelysin (MMP3/Transin) degrades several matrix components including the core proteins of proteoglycans, laminin and non-helical regions of collagens.

Host Species: Mouse

Clone: SL-1 ID3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)

Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)

Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

MMP-3 (Stromelysin-1) Ab-1
Monoclonal
200μg/mL
Concentrated
APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
1mL
Proteases And Inhibitors
Primary
Human
Immunofluorescence, Western Blot
SL-1 ID3
Biotin
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Videos
SDS
Documentos