Learn More
Epredia™ Lab Vision™ Mesothelin Ab-1 Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelin Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1320-S0
Detalles adicionales : Peso : 0.00170kg
Descripción
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, that is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.Host Species: Mouse
Clone: 5B2
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule
Molecular Weight: 40kDa
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Mesothelin Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
5B2 | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Human | |
IgG1 |