missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Mast Cell Tryptase Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Epredia™ Mast Cell Tryptase Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia™ MS-1216-PABX

Detalles adicionales : Peso : 0.00180kg

 Ver más versiones de este producto

Código de producto. 12669296

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of tryptase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. In vitro studies indicate tryptase can cleave C3 to form C3a anaphylatoxin, inactivate fibrinogen as a coaguable substrate for thrombin and activate latent collagenase. Human mast cell tryptase has considerable potential as a marker for mast cell activation as well as being a mediator of inflammation.

Host Species: Mouse

Clone: AA1

Isotype: IgG1

Species Reactivity: Human, Monkey, Dog and Cat. Others not known.

Epitope: Not determined

Immunogen: Human mast cell tryptase purified from human lung tissues

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Mast Cell Tryptase Ab-2
Monoclonal
1mg/mL
Mouse
200 μL
Enzyme
Canine, Feline, Human, Non-human Primate
IgG1
Immunohistochemistry (Paraffin), Western Blot
AA1
Unconjugated
Human mast cell tryptase purified from human lung tissues
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales