missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Mammaglobin Mouse Monoclonal Antibody, BSA and Azide

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mammaglobin, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1919-A0

Detalles adicionales : Peso : 0.00170kg

 Ver más versiones de este producto

Código de producto. 12670467

  • 302.00€ / 100 microlitros

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Beschreibung

Beschreibung

Mammaglobin, a member of the uteroglobin family, is a 10kDa glycoprotein. Mammaglobin mRNA expression is breast tissue specific. It is overexpressed in human breast cancer. The use of Mammaglobin RT-PCR has been demonstrated to detect disseminated breast cancer. In a study to evaluate the frequency and specificity of mammaglobin, 80% of all primary and metastatic breast tumors tested were immunopositive irrespective of tumor grade, while mammaglobin mRNA was detected in 60% of the peripheral stem cell collections from breast cancer patients.

Host Species: Mouse

Clone: 304-1A5

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant full-length protein

Molecular Weight: 10kDa

Positive Control: Breast carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Spezifikation

Spezifikation

Mammaglobin
Monoclonal
Unconjugated
Murine
IgG1
ASR
Primary
Monoclonal
Immunohistochemistry (Paraffin)
304-1A5
Concentrated
Recombinant full-length protein
100μL
Cancer and Tumor Biology
Primary
Human
Videos
Sicherheitsdatenblatt (SDS)
missing translation for 'documents'
Product Certifications