missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ LCK (p56lck) Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Recommended for Western Blotting, Immunoprecipitation (Denatured verified) and Kinase Assays, Epredia™ LCK (p56lck) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results. 

Marca:  Epredia™ MS-164-P

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12690027

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »


Descripción

Descripción

p56lck is a membrane-associated non-receptor protein tyrosine kinase from the Src-family of kinases. The phosphorylation status and the activity of p56lck is regulated by the CD45 tyrosine protein phosphatase. p56lck plays a critical role in T cell development and activation. p56lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas.

Host Species: Mouse

Clone: 3A5

Isotype: IgG2b/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1-225

Immunogen: Recombinant protein corresponding to aa 1-225 of murine p56lck protein

Molecular Weight: 56kDa

Positive Control: Jurkat cells

Cellular Localization: Cell membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

LCK (p56superscriptlcksuperscript) Ab-1
Monoclonal
200μg/mL
Concentrated
Recombinant protein corresponding to aa 1-225 of murine p56superscriptlcksuperscript protein
1mL
Cancer and Tumor Biology
Primary
Human, Murine, Rat
Immunoprecipitation, Western Blot
3A5
Unconjugated
Murine
IgG2b kappa
RUO
Primary
Monoclonal
Videos
SDS
Documentos