missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Lambda Light Chain Ab-4 (Cocktail) Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia™ MS-1158-P

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12669136

  • 296.00€ / 1 ml

Envío estimado: 01-07-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: LAM03 + HP6054

Isotype: IgG1/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-4 cocktail is especially designed for sensitive detection of lambda light chain of immunoglobulin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Lambda Light Chain Ab-4 (Cocktail)
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Immunohistochemistry (Paraffin)
LAM03 + HP6054
Unconjugated
Concentrated
Purified human lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
1mL
Hematopoietic
Primary
Human
Videos
SDS
Documentos
Certificados