missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ HER-2/c-erbB-2/neu Ab-8 Mouse Monoclonal Antibody, 200µg/mL, biotinylated, BSA and azide

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia™ MS-325-P

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12681017

  • 525.00€ / 1 ml

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

c-erbB-2 is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein. The c-erbB-2 protein is overexpressed in a variety of carcinomas especially those of breast and ovary.

Host Species: Mouse

Clone: e2-4001

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Cytoplasmic domain

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunoprecipitation (Native and Denatured)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

HER-2/c-erbB-2/neu Ab-8
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG1
RUO
Primary
Monoclonal
Immunoprecipitation
e2-4001
Unconjugated
Concentrated
Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein
1mL
Oncogenesis
Primary
Human, Murine, Rat
Videos
SDS
Documentos
Certificados