missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Gastrin Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Obtain accurate, reproduicble results in immunohistochemistry experiments with Epredia™ Gastrin Ab-1, Rabbit Polyclonal Antibody.

Marca:  Epredia™ RB-1459-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12672917

  • 373.00€ / 7 ml

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Gastrin, a polypeptide hormone, occurs naturally in three forms: gastrin-14, gastrin-17 and gastrin-34.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Immunogen: A synthetic peptide from human gastrin

Positive Control: Stomach

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin )
Especificaciones

Especificaciones

Gastrin Ab-1
Polyclonal
Unconjugated
Rabbit
IgG
IVD
Primary
Polyclonal
Immunohistochemistry (Paraffin)
Polyclonal
Prediluted
A Synthetic peptide from human gastrin
7mL
Steroids Hormones and Growth Factors
Primary
Human
Videos
SDS
Documentos
Certificados