Learn More
Filaggrin Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Filaggrin Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-449-P1ABX
Detalles adicionales : Peso : 1.00000kg
Descripción
Filaggrin is expressed only in well-differentiated, keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.Host Species: Mouse
Clone: FLG01
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant filaggrin protein
Molecular Weight: 38 kDa (Filaggrin) and 400 kDa (Profilaggrin)
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Filaggrin Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cytoskeletal Protein | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
FLG01 | |
Unconjugated | |
Recombinant filaggrin protein | |
RUO | |
Primary | |
Concentrated |