Learn More
Epithelial Specific Antigen/Ep-CAM Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-144-PABX
Detalles adicionales : Peso : 1.00000kg
Descripción
EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.Host Species: Mouse
Clone: VU-1D9
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Small cell lung carcinoma cells
Molecular Weight: 40-43 kDa
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Epithelial Specific Antigen/Ep-CAM Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Cancer and Tumor Biology | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
VU-1D9 | |
Unconjugated | |
Small cell lung carcinoma cells | |
IVD | |
Primary | |
Concentrated |