missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 5 Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 5, Rabbit Monoclonal Antibody.

Marca:  Epredia™ RM-2106-S

Detalles adicionales : Peso : 0.00250kg

 Ver más versiones de este producto

Código de producto. 12673617

  • 1170.00€ / 1 ml
Fecha estimada de envío: 06-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Twenty human cytokeratins are divided into acidic (pI<5.7) and basic (pI>6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of cytokeratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of cytokeratins as markers in cancer research and tumor identification. Point mutations in cytokeratin-5 gene may cause Epidermolysis Bullosa Simplex and Dowling-Degos disease. cytokeratin-5 is expressed in most epithelioid and biphasic mesotheliomas, thymoma, squamous and other carcinomas. It is related to cytokeratin-6.

Host Species: Rabbit

Clone: EP1601Y

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues on the C-terminus of human CK5

Molecular Weight: 59 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

Cytokeratin 5
Monoclonal
Unconjugated
A synthetic peptide corresponding to residues on the C-term of human CK5
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
EP1601Y
Rabbit
1 mL
Cancer and Tumor Biology
Human
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales