missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Collagen II Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Ensure accurate, reproducible results in western blotting and immunofluorescence experiments with Epredia™ Collagen II Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-234-P1

Detalles adicionales : Peso : 0.00190kg

 Ver más versiones de este producto

Código de producto. 12680647

  • 332.00€ / 500 microlitros

Fecha estimada de envío
21-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Host Species: Mouse

Clone: 5B2.5

Isotype: IgG2a/κ

Species Reactivity: Human, Chicken and Mouse. Others not known.

Epitope: GGFDEK, slightly preceding the lysine residue involved in crosslinking. Two terminal glycine residues constitute a key structural element of the epitope. The epitope is lost after digestion of lathyritic type II collagen with recombinant stromelysin.

Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum

Positive Control: Cartilage

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Collagen II Ab-1
Monoclonal
200μg/mL
Concentrated
A purified preparation of lathyritic type II collagen from embryonic chicken sternum
500μL
Extracellular Matrix
Primary
Chicken, Human, Murine
Immunofluorescence, Western Blot
5B2.5
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Videos
SDS
Documentos