Learn More
Collagen II Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in western blotting and immunofluorescence experiments with Epredia™ Collagen II Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-234-P0
Detalles adicionales : Peso : 1.00000kg
Descripción
Host Species: Mouse
Clone: 5B2.5
Isotype: IgG2a/κ
Species Reactivity: Human, Chicken and Mouse. Others not known.
Epitope: GGFDEK, slightly preceding the lysine residue involved in crosslinking. Two terminal glycine residues constitute a key structural element of the epitope. The epitope is lost after digestion of lathyritic type II collagen with recombinant stromelysin.
Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Positive Control: Cartilage
Cellular Localization: Cartilage matrix
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Especificaciones
Collagen II Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Extracellular Matrix | |
Chicken, Human, Mouse | |
IgG2a κ |
Immunofluorescence, Western Blot | |
5B2.5 | |
Unconjugated | |
A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
RUO | |
Primary | |
Concentrated |