Learn More
Epredia™ Lab Vision™ Chorionic Gonadotropin Human/hCG Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1065-P1
Detalles adicionales : Peso : 1.00000kg
Descripción
Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.Host Species: Mouse
Clone: CG04 + CG05
Isotype: IgG2a/κ + IgG1/κ
Species Reactivity: Human. Others not known
Immunogen: Purified β-subunit of hCG
Molecular Weight: 22 kDa (β-hCG)
Positive Control: Placenta or chorioca
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not optimum)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Chorionic Gonadotropin, Human/hCG Ab-5 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Steroids Hormones and Growth Factors | |
Human | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
CG04 + CG05 | |
Unconjugated | |
Purified Beta-subunit of HCG | |
RUO | |
Primary | |
Concentrated |