missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Chorionic Gonadotropin Human/hCG Ab-5 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1065-PABX

Detalles adicionales : Peso : 0.00180kg

 Ver más versiones de este producto

Código de producto. 12688796

  • 341.00€ / 200 microlitros

Envío estimado: 01-07-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Mouse

Clone: CG04 + CG05

Isotype: IgG2a/κ + IgG1/κ

Species Reactivity: Human. Others not known

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa (β-hCG)

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Chorionic Gonadotropin, Human/hCG Ab-5
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG1 kappa
RUO
Primary
Monoclonal
Immunohistochemistry (Paraffin)
CG04 + CG05
Unconjugated
Concentrated
Purified Beta-subunit of HCG
200μL
Steroids Hormones and Growth Factors
Primary
Human
Videos
SDS
Documentos
Certificados