missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD42b (GPIb) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1174-P

Detalles adicionales : Peso : 0.00250kg

 Ver más versiones de este producto

Código de producto. 12689206

  • 725.00€ / 1 ml
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The CD42b glycoprotein, also known as GPIb, is a cofactor of ristocetin-induced aggregation and is involved in the binding of platelets to blood vessel walls. The CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. The absence of CD42b antigen on platelets may indicate Bernard-Soulier disease.

Host Species: Mouse

Clone: 42C01

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant CD42b.

Molecular Weight: 135-145 kDa (α-chain) and 22-25 kDa (β-chain)

Positive Control: Tonsil

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

CD42b Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
Hematopoietic
Human
IgG1
Immunohistochemistry (Paraffin)
42C01
Unconjugated
Recombinant CD42b
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales