missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD1a Ab-5 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry applications with Epredia™ CD1a Ab-5, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1856-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12680317

  • 288.00€ / 7 ml

Envío estimado: 01-07-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

At least five CD1 genes (CD1a, b, c, d, and e) have been identified. CD1 is expressed on cortical thymocytes, Langerhans cells, and dendritic cells. It is absent on mature peripheral blood T cells but intracytoplasmic expression is detected on activated T lymphocytes. CD1 proteins have been demonstrated to restrict T-cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation.

Host Species: Mouse

Clone: O10

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Molecular Weight: 49kDa

Positive Control: Tonsil

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

CD1a Ab-5
Monoclonal
Unconjugated
Prediluted
IgG2a kappa
IVD
Primary
Monoclonal
Immunohistochemistry (Paraffin)
O10
Ready-to-use for IHC (UVLP)
Murine
7mL
Hematopoietic
Primary
Human
Videos
SDS
Documentos
Certificados