missing translation for 'onlineSavingsMsg'
Learn More

Calretinin Rabbit Polyclonal Antibody, Epredia™

Stain formalin-fixed, paraffin-embedded tissues using Epredia™ Calretinin, Rabbit Polyclonal Antibody.

Marca:  Epredia RB-9002-P0

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 12673277

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length mouse calretinin protein

Molecular Weight: 31.5 kDa

Positive Control: Mesothelioma

Cellular Localization: Nuclear and Cytoplasmic

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

Calretinin
Polyclonal
Unconjugated
Recombinant full-length mouse calretinin protein
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
200µg/mL
Rabbit
100 μL
Oncogenesis
Human, Mouse, Rat
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales