missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calponin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Calponin Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1168-P1

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12679176

  • 331.00€ / 500 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Calponin, a calmodulin, binds tropomyosin and F-actin and is thought to be involved in the regulation of smooth muscle contraction. The expression of calponin is restricted to smooth muscle cells and is a marker of the differentiated contractile phenotype of developing smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions.

Host Species: Mouse

Clone: CALP

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not tested.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 34 kDa

Positive Control: Myoepithelial cells in breast ducts

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

Calponin Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Muscle Biology
Human, Rat
IgG2a κ
Immunohistochemistry (Paraffin)
CALP
Unconjugated
Crude human uterus extract
IVD
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales