missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Androgen Receptor Ab-1 Mouse Monoclonal Antibody, 200µg/mL, biotinylated, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and gel supershift experiments.

Marca:  Epredia™ MS-443-P0

Detalles adicionales : Peso : 4.98950kg

 Ver más versiones de este producto

Código de producto. 12681427

  • 124.00€ / 100 microlitros

Envío estimado: 02-07-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The expression of AR is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Mouse

Clone: AR 441

Isotype: IgG1

Species Reactivity: Human and Dog. Does not react with mouse. Others not tested.

Epitope: aa 299-315

Immunogen: A synthetic peptide from human AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

Ab-1 is excellent for staining of formalin/paraffin tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Androgen Receptor Ab-1
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG1
RUO
Primary
Monoclonal
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
AR 441
Unconjugated
Concentrated
A synthetic peptide from human AR
100μL
Steroids Hormones and Growth Factors
Primary
Canine, Human
Videos
SDS
Documentos
Certificados