missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody with BSA and azide

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

Marca:  Epredia™ RB-9023-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12673337

  • 164.00€ / 7 ml

Fecha estimada de envío
21-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recognizes APP (b-APP770), MW 110kDa. Predicted to recognize Β-APP695 and Β-APP751. Amyloid precursor protein (APP) and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide corresponding to N-terminal of human APP

Molecular Weight: 110kDa

Positive Control: Brain

Cellular Localization: Cytoplasmic and extracellular

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Especificaciones

Especificaciones

Amyloid Precursor Protein (APP)
Polyclonal
Prediluted
A synthetic peptide corresponding to N-terminal of human APP
7mL
Neuroscience
Primary
Human, Murine, Rat
Western Blot
Unconjugated
Rabbit
IgG
IVD
Primary
Polyclonal
Videos
SDS
Documentos