missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ ALK (Anaplastic Lymphoma Kinase)/p80 Ab-1 Mouse Monoclonal Antibody, 7mL

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ ALK (Anaplastic Lymphoma Kinase)/p80 Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1104-S1

Detalles adicionales : Peso : 0.00190kg

 Ver más versiones de este producto

Código de producto. 12668966

  • 341.00€ / 500 microlitros

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

ALK/p80 is a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in a third of large cell lymphomas.

Host Species: Mouse

Clone: 5A4

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: aa 419-520 (tyrosine kinase domain)

Immunogen: Recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript (419-520aa)

Molecular Weight: 80kDa

Positive Control: Anaplastic lymphoma

Cellular Localization: Cytoplasmic and/or nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not recommended)
Especificaciones

Especificaciones

ALK (Anaplastic Lymphoma Kinase)/p80 Ab-1
Monoclonal
Unconjugated
Murine
IgG1
IVD
Primary
Monoclonal
Immunohistochemistry (Paraffin)
5A4
Concentrated
Recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript (419-520aa)
500μL
Oncogenesis
Primary
Human
Videos
SDS
Documentos
Certificados