missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Actin, Skeletal Muscle, Ab-2 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin Skeletal Muscle Ab-2 Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-185-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12680337

  • 160.00€ / 7 ml

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Host Species: Mouse

Clone: 5C5.F8.C7 (α-Sr-1)

Isotype: IgM/κ

Species Reactivity: Human, Cow, Rabbit, Sheep, Rat, Guinea Pig and Frog. Others not known.

Epitope: N-terminal decapeptide

Immunogen: N-Terminal decapeptide of α skeletal muscle isoform of actin; acetylated at the N-terminus

Positive Control: Skeletal or Cardiac muscle, or Rhabdomyosarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Actin, Skeletal Muscle Ab-2
Monoclonal
Unconjugated
Prediluted
Immunogen: N-Terminal decapeptide of alpha skeletal muscle isoform of actin; acetylated at the N-terminus
7mL
Muscle Biology
Primary
Bovine, Frog, Guinea Pig, Human, Rabbit, Rat, Sheep
Immunofluorescence, Immunohistochemistry (Paraffin)
5C5.F8.C7; or alpha-Sr-1
Ready-to-use for IHC (UVLP)
Murine
IgM kappa
RUO
Primary
Monoclonal
Videos
SDS
Documentos
Certificados