Learn More
L-Threoninol, TRC
Product Code. 30454923
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30454923 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 30454923 Supplier Toronto Research Chemicals Supplier No. TRCT405578100MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Threoninol
CAS 3228-51-1
Molecular Formula C4H11NO2
Quantity 100 mg
InChI Formula InChI=1S/C4H11NO2/c1-3(7)4(5)2-6/h3-4,6-7H,2,5H2,1H3/t3-,4-/m1/s1
SMILES C[C@@H](O)[C@H](N)CO
IUPAC Name (2R,3R)-2-aminobutane-1,3-diol
Molecular Weight (g/mol) 105.14
Formula Weight 105.079