Learn More
L-Lysine hydrochloride, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Catalog Number p-7082824
Click to view available options
Cantidad:
1 kg
5 kg
10 kg
Envase:
Balde de polietileno
Frasco de plástico
Tambor de fibra
657-27-2
C6H15ClN2O2
182.648
MFCD00064564
BVHLGVCQOALMSV-JEDNCBNOSA-N
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
69568
CHEBI:53633
C(CCN)CC(C(=O)O)N.Cl
657-27-2
C6H15ClN2O2
182.648
MFCD00064564
BVHLGVCQOALMSV-JEDNCBNOSA-N
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
69568
CHEBI:53633
C(CCN)CC(C(=O)O)N.Cl
Nombre del producto químico o material L-Lysine hydrochloride
Punto de fusión ∼260°C (decomposition)
Notas del porcentaje del ensayo Base seca
Fórmula lineal NH2(CH2)4CH(NH2)COOH.HCl
Beilstein 3563889
Envase Frasco de plástico
Nombre IUPAC (2S)-2,6-diaminohexanoic acid hydrochloride
Formula Weight (peso de la fórmula) 182.65
Porcentaje de pureza ≥98.5%
Cantidad 1 kg
Forma física Polvo cristalino