Learn More
L-(+)-Lactic Acid (Free Acid), Fisher BioReagents
Product Code. 11321196
Change view
Click to view available options
Amber Glass; 5g
Product Code.
11321196 Amber Glass; 5g
1 options
This item is not returnable. View return policy
Product Code. 11321196 Supplier Fisher BioReagents Supplier No. BP26615
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
L-(+)-Lactic Acid is a biochemical that contains varying amounts of intermolecular esterification products.
TRUSTED_SUSTAINABILITY
Specifications
pH 1.75
CAS 79-33-4
Melting Point 53°C
Boiling Point 119°C
Color White
Assay Percent Range ≥98 %
Molecular Formula C3H6O3
Merck Index 15, 5385
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
SMILES CC(C(=O)O)O
IUPAC Name (2S)-2-hydroxypropanoic acid
Molecular Weight (g/mol) 90.078
PubChem CID 107689
ChEBI CHEBI:422
Formula Weight 90.08
Percent Purity 98% min.
Density 1.20g/cm³
Quantity 5 g
Infrared Spectrum Conforms to reference
Packaging Amber Glass
Physical Form Powder/Solid
Vapor Pressure 0.0813mmHg at 25°C
Chemical Name or Material L-(+)-Lactic Acid, Free acid