Learn More
L-beta-prolinol, 95 %, Thermo Scientific Chemicals
Catalog Number p-7038279
Click to view available options
Cantidad:
250 mg
1 g
110013-19-9
C5H11NO
101.149
MFCD09260722
QOTUIIJRVXKSJU-YFKPBYRVSA-N
s-pyrrolidin-3-ylmethanol, 3s-pyrrolidin-3-ylmethanol, l-beta-prolinol, s-3-pyrrolidin-methanol, 3-pyrrolidinemethanol, 3s, 3-pyrrolidinemethanol, s, 3s-pyrrolidin-3-yl methanol, s-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine
7446911
[(3S)-pirrolidina-3-il]metanol
C1CNCC1CO
110013-19-9
C5H11NO
101.149
MFCD09260722
QOTUIIJRVXKSJU-YFKPBYRVSA-N
s-pyrrolidin-3-ylmethanol, 3s-pyrrolidin-3-ylmethanol, l-beta-prolinol, s-3-pyrrolidin-methanol, 3-pyrrolidinemethanol, 3s, 3-pyrrolidinemethanol, s, 3s-pyrrolidin-3-yl methanol, s-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine
7446911
[(3S)-pirrolidina-3-il]metanol
C1CNCC1CO
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Se utiliza como intermediario farmacéutico.
Solubilidad
Ligeramente soluble en agua
Notas
Higroscópico. Almacenar a temperatura ambiente.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material L-beta-Prolinol
Densidad 0.979
Punto de fusión 174°C to 176°C
Cantidad 250 mg
Sensibilidad Hygroscopic
Información de solubilidad Slightly soluble in water
Formula Weight (peso de la fórmula) 101.15
Porcentaje de pureza 95%