missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRT4 (Human) Recombinant Protein
Human KRT4 full-length ORF ( AAH42174.1, 1 a.a. - 534 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003851-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MIARQQCVRGGPRGFSCGSAIVGGGKRGAFSSVSMSGGAGRCSSGGFGSRSLYNLRGNKSISMSVAGSRQGACFGGAGGFGTGGFGAGGFGAGFGTGGFGGGFGGSFSGKGGPGFPVCPAGGIQEVTINQSLLTPLHVEIDPEIQKVRTEEREQIKLLNNKFASFIDKVQFLEQQNKVLETKWNLLQQQTTTTSSKNLEPLFETYLSVLRKQLDTLGNDKGRLQSELKTMQDSVEDFKTKYEEEINKRTAAENDFVVLKKDVDAAYLNKVELEAKVDSLNDEINFLKVLYDAELSQMQTHVSDTSVVLSMDNNRNLDLDSIIAEVRAQYEEIAQRSKAEAEALYQTKVQQLQISVDQHGDNLKNTKSEIAELNRMIQRLRAEIENIKKQCQTLQVSVADAEQRGENALKDAHSKRVELEAALQQAKEELARMLREYQELMSVKLALDIEIATYRKLLEGEEYRMSGECQSAVSISVVSGSTSTGGISGGLGSGSGFGLSSGFGSGSGSGFGFGGSVSGSSSSKIISTTTLNKRR
Especificaciones
AAH42174.1 | |
keratin 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRT4 | |
GST |
3851 | |
Wheat germ expression system | |
25 μg | |
CK4, CYK4, FLJ31692, K4 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |