missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13963-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC) which consists of 60-100 proteins and is probably 120 million daltons in molecular size. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion; larger molecules are transported by an active process. Most nuclear proteins contain short basic amino acid sequences known as nuclear localization signals (NLSs). KPNA5 protein belongs to the importin alpha protein family and is thought to be involved in NLS-dependent protein import into the nucleus.Especificaciones
KPNA5 | |
Polyclonal | |
Unconjugated | |
KPNA5 | |
Importin subunit alpha 6, IPOA6, Karyopherin subunit alpha 5, KPNA5, SRP6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16650, 294392, 3841 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15131, O35345, Q56R16 | |
KPNA5, Kpna6 | |
KPNA5 Fusion Protein Ag4920 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |