missing translation for 'onlineSavingsMsg'
Learn More

KLRG1 Syrian Hamster anti-Mouse, Super Bright 780, Clone: 2F1, eBioscience™

Syrian Hamster Monoclonal Antibody

Marca:  Invitrogen 78-5893-80

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15934184

  • 135.00€ / 25 microgramos


Fecha de envío estimada
del proveedor 22-11-2019
Añadir a la cesta

Descripción

Descripción

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 780 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 780 nm. We recommend using a 780/60 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product No. 01-2222-42) as compared to single-color stained cells. In such circumstances, we would recommend setting compensation with cells. We have also observed this in some experiments using AbC Total Antibody Compensation beads (Product No. A10497). Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightnessand FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 780 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. Functionally, KLRG1 is a lymphocyte that is involved in the lysis of certain tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.
Especificaciones

Especificaciones

KLRG1
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
O88713
Klrg1
IgG
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
50928
Flow Cytometry
2F1
Super Bright 780
Klrg1
Liquid
Syrian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos
Documentos