missing translation for 'onlineSavingsMsg'
Learn More

KLK15 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 27515-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16825675

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. In prostate cancer, this gene has increased expression, which indicates its possible use as a diagnostic or prognostic marker for prostate cancer. The gene contains multiple polyadenylation sites and alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications

Specifications

KLK15
Polyclonal
Unconjugated
KLK15
ACO, ACO protease, HSRNASPH, Kallikrein 15, KLK15
Rabbit
Antigen Affinity Chromatography
RUO
55554
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H2R5
KLK15
KLK15 Fusion Protein Ag26232
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers