missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27515-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. In prostate cancer, this gene has increased expression, which indicates its possible use as a diagnostic or prognostic marker for prostate cancer. The gene contains multiple polyadenylation sites and alternative splicing results in multiple transcript variants encoding distinct isoforms.Especificaciones
KLK15 | |
Polyclonal | |
Unconjugated | |
KLK15 | |
ACO, ACO protease, HSRNASPH, Kallikrein 15, KLK15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55554 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H2R5 | |
KLK15 | |
KLK15 Fusion Protein Ag26232 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |