missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17117-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Kelch-like protein 11, Subcellular location Secreted.Especificaciones
KLHL11 | |
Polyclonal | |
Unconjugated | |
KLHL11 | |
kelch like 11 (Drosophila), Kelch like protein 11, KLHL11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217194, 287706, 55175 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CE33, Q9NVR0 | |
Klhl11 | |
KLHL11 Fusion Protein Ag10805 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |