missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27277-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
KLHDC1 contains 6 Kelch repeats and is widely expressed, with high levels in skeletal muscle, pancreas and liver. KLHDC1 and KLHDC2 have differential localization and activity in cultured mammalian cells. The exact function of KLHDC1 remains unknown.Especificaciones
KLHDC1 | |
Polyclonal | |
Unconjugated | |
KLHDC1 | |
kelch domain containing 1, KLHDC1, MST025 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122773, 271005, 314190 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80YG3, Q8N7A1 | |
Klhdc1 | |
KLHDC1 Fusion Protein Ag23704 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |