missing translation for 'onlineSavingsMsg'
Learn More

KLF9 Recombinant Rabbit Monoclonal Antibody (5H16L7), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marca:  Invitrogen 701888

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15662025

  • 470.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is predicted to react with Bovine, Pig and Monkey. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

KLF9 Recombinant Monoclonal antibody specifically detects KLF9 in Human, Mouse, Rat samples. It is validated for ChIP Assay, Western Blot.
Especificaciones

Especificaciones

KLF9
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q13886, O35739, Q01713
KLF9
Peptides corresponding to Human KLF9 (aa 34-49, 85-98)
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
ChIP Assay, Western Blot
5H16L7
Unconjugated
KLF9
2310051E17Rik; AA589643; basic transcription element binding protein 1; basic transcription element-binding protein 1; BTEB; BTEB1; BTEB-1; BTE-binding protein 1; ENSMUSG00000055440; GC-box-binding protein 1; Gm9971; KLF9; Krueppel-like factor 9; Kruppel like factor 9; Kruppel-like factor 9; transcription factor BTEB1
Rabbit
Protein A
RUO
117560, 16601, 687
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales