Learn More
Kimberly-Clark™ KLEENGUARD™ A50 Breathable Splash and Particle Protection Trousers
Specially designed for comfort, safety and strength. Kimberly-Clark™ KLEENGUARD™ A50 Breathable Splash and Particle Protection Trousers is designed for comfort, safety and general protection against larger particles, radioactive dust and chemical splashes.
Marca:  Kimberly-Clark™ 99510
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12681446
  • 126.00€ / paquete de 15
Fecha estimada de envío
17-12-2019
Añadir a la cesta
Descripción
Descripción
  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
  • Single use and breathable protective trousers designed for maximum comfort and safety, providing anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • Durable SMS fabric with additional treatment for increased protection
  • Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
  • Silicone-free, ultra-low lint and treated to be antistatic - to EN 1149-5
  • Ideal for protecting against powders, larger fibres and protection against a wide range of chemicals - reducing contamination in critical areas
  • Suitable for use  in pharmaceutical industries, manufacturing, utilities, electronics, agriculture and paint spraying areas
Especificaciones
Especificaciones
White
Larger particles, radioactive dust and chemical splashes.
Double-layer fabric
Silicone Free
Large
Providing high levels of protection against larger particles, radioactive dust and chemical splash
Breathable Splash and Particle Protection Trousers