Learn More
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves are an ambidextrous glove that provides protection against chemical splash in a wide range of applications.
Marca:  Kimberly-Clark™ 90094
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12638835
  • 313.00€ / paquete de 2250
Fecha estimada de envío
10-01-2020
Añadir a la cesta
Descripción
Descripción
  • PPE Category 3 protection
  • For increased protection against chemical splashes of several chemicals.
  • Provides exceptional touch sensitivity and textured fingertips to improve grip and the handling of small objects
  • AQL 0.65 for pinholes guarantees quality of production process, waste reduction and better protection
  • Independently accredited
  • Suitable for food handling
  • Beaded cuff for added strength and ease of donning
  • Latex Free
  • Powder free minimises srying effects of pwder and skin allergies
  • Ambidextrous
  • Portable dispenser box for mobility and easy access
Especificaciones
Especificaciones
PPE Category III
Beaded/Rolled
G20
2250Pack
0.06 mm
Textured
Ideal for providing PPE category 3 protection against chemical splash
X-Large
Atlantic Green
Gloves