missing translation for 'onlineSavingsMsg'
Learn More

KIR2DS2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-82102DL594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214527

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

KIR2DS2 Polyclonal antibody specifically detects KIR2DS2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Especificaciones

Especificaciones

KIR2DS2
Polyclonal
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
Peptide Affinity-purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunofluorescence
DyLight 594
183ActI, CD158b, CD158J, cl-49, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2, NKAT5, p58 KIR
KIR2DS2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human KIR2DS2. The immunogen is located within the last 50 amino acids of KIR2DS2. Amino Acid Squence: EPAGNRTVNSEDSDEQDH
0.1 mL
Immunology
100132285
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales