missing translation for 'onlineSavingsMsg'
Learn More

KIR2DL4/CD158d Antibody (2H6), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-04312C

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213474

  • 521.58€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

KIR2DL4/CD158d
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
KIR2DL4
Recombinant human KIR2DL4 (44-202 aa) purified from E. coli
0.1 mL
Immunology
3805
Human
Purified
Western Blot, ELISA
2H6
Western Blot, ELISA
103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor
Mouse
Protein A purified
RUO
Primary
Anti-human KIR2DL4 mAb, clone 2H6, is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c immunized with recombinant human KIR2DL4 protein.
Store at 4C in the dark.
IgG2b κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales