Learn More
Kinematica Polytron™ Homogenizer Generator Tool
Product Code. 11837194 Shop All Kinematica Products
Change view
Click to view available options
For Use With (Equipment):
Polytron™ generators
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11837194 Polytron™ generators Each
1 options
This item is not returnable. View return policy
Product Code. 11837194 Supplier Kinematica Supplier No. 11090010
  • ONLINE EXCLUSIVE PRICE
    221.00€
    169.00€ / paquete de 1
    Ahorro 52.00€
    23% Descuento
Fecha estimada de envoi: 04-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Designed for quick and easy service of Polytron™ generators.
  • Fits all Polytron™ generators between 5 and 60 mm
  • Precision made by skilled Swiss craftsmen
Specifications
Description Homogenizer tool
For Use With (Application) Changing rotors on homogenizers
For Use With (Equipment) Polytron™ generators